1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid

Lieferant: BIONET RESEARCH

2R-1304.500 2R-1304.1 2R-1304.5 2R-1304.10
BIOT2R-1304.500EA 0 EUR
BIOT2R-1304.500 BIOT2R-1304.1 BIOT2R-1304.5 BIOT2R-1304.10
1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid
1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäurehydrazid
Formel: C₁₃H₁₉N₃O₃S
MDL: MFCD00759493
CAS-Nummer: 324531-31-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR