1-((4-Nitrophenyl)sulfonyl)-4-piperidincarbonsäure

Lieferant: BIONET RESEARCH

4R-1331.500 4R-1331.1 4R-1331.5 4R-1331.10
BIOT4R-1331.500EA 0 EUR
BIOT4R-1331.500 BIOT4R-1331.1 BIOT4R-1331.5 BIOT4R-1331.10
1-((4-Nitrophenyl)sulfonyl)-4-piperidincarbonsäure
1-((4-Nitrophenyl)sulfonyl)-4-piperidincarbonsäure
Formel: C₁₂H₁₄N₂O₆S
MDL: MFCD01045658
CAS-Nummer: 331828-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR