1-((4-Methoxyphenyl)sulfonyl)-4-piperidincarbonsäure
Lieferant: BIONET RESEARCH
Synonyme:
1-((4-Methoxyphenyl)sulphonyl)piperidine-4-carboxylic acid
4R-1334.500
4R-1334.1
4R-1334.5
4R-1334.10
BIOT4R-1334.500EA
0
EUR
BIOT4R-1334.500
BIOT4R-1334.1
BIOT4R-1334.5
BIOT4R-1334.10
1-((4-Methoxyphenyl)sulfonyl)-4-piperidincarbonsäure
1-((4-Methoxyphenyl)sulfonyl)-4-piperidincarbonsäure
Formel:
C₁₃H₁₇NO₅S Molecular Weight: 299.340 g/mol |
MDL:
MFCD02331441 CAS-Nummer: 385398-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...