1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 1-Tosylpiperidine-4-carboxylic acid

4R-1329.500 4R-1329.1 4R-1329.5 4R-1329.10
BIOT4R-1329.500EA 0 EUR
BIOT4R-1329.500 BIOT4R-1329.1 BIOT4R-1329.5 BIOT4R-1329.10
1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäure
1-((4-Methylphenyl)sulfonyl)-4-piperidincarbonsäure
Formel: C₁₃H₁₇NO₄S
Molecular Weight: 283,3400 g/mol
MDL: MFCD00723645
CAS-Nummer: 147636-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR