1H-Benzimidazol-5-boronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzimidazole, Benzimidazole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole

708879-250MG 708879-1G
SIAL708879-250MGEA 47.3 EUR
SIAL708879-250MG SIAL708879-1G
1H-Benzimidazol-5-boronsäurepinakolester, Sigma-Aldrich®
1H-Benzimidazol-5-boronsäurepinakolester
Formel: C₁₃H₁₇BN₂O₂
Molecular Weight: 244.10 g/mol
MDL: MFCD11054041
CAS-Nummer: 1007206-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR