5-Amino-2-mercaptobenzimidazol

Lieferant: Thermo Scientific

Synonyme: 5-Amino-1H-benzoimidazole-2-thiol

AC34641DA AC34641EA AC34641EE
MAYBAC34641DAEA 0 EUR
MAYBAC34641DA MAYBAC34641EA MAYBAC34641EE
5-Amino-2-mercaptobenzimidazol
5-Amino-2-mercaptobenzimidazole
Formel: C₇H₇N₃S
MDL: MFCD00022671
CAS-Nummer: 2818-66-8
EINECS: 220-574-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR