5-(Trifluormethoxy)isatin ≥98%

Lieferant: Thermo Scientific

Synonyme: 5-(Trifluoromethoxy)indoline-2,3-dione

B25494.03 B25494.06
B25494.03EA 33.1 EUR
B25494.03 B25494.06
5-(Trifluormethoxy)isatin ≥98%
5-(Trifluormethoxy)isatin

Purity: 98% CAS No.: 169037-23-4 Molecular Formula: C9H4F3NO2 Formula Weight: 231.13 Melting Point: 170-172° MDL No.: MFCD00192524

Formel: C₉H₄F₃NO₃
Molecular Weight: 231,13 g/mol
Schmelzpunkt: 170…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192524
CAS-Nummer: 169037-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR