5-(Trifluormethoxy)isatin

Lieferant: COMBI-BLOCKS

Synonyme: 5-(Trifluoromethoxy)indoline-2,3-dione

IN-0136-5G IN-0136-1G IN-0136-25G
COBBIN-0136-5GEA 137 EUR
COBBIN-0136-5G COBBIN-0136-1G COBBIN-0136-25G
5-(Trifluormethoxy)isatin
5-(Trifluormethoxy)isatin
Formel: C₉H₄F₃NO₃
Molecular Weight: 231,13 g/mol
MDL: MFCD00192524
CAS-Nummer: 169037-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR