6-(Trifluormethyl)isatin

Lieferant: COMBI-BLOCKS

IN-0179-5G IN-0179-1G
COBBIN-0179-5GEA 0 EUR
COBBIN-0179-5G COBBIN-0179-1G
6-(Trifluormethyl)isatin
6-(Trifluormethyl)isatin
Formel: C₉H₄F₃NO₂
MDL: MFCD08457887
CAS-Nummer: 343-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR