3,4-(Methylendioxy)benzylamin
Lieferant: FLUOROCHEM
Synonyme:
Piperonylamine, Benzo[d][1,3]dioxol-5-ylmethanamine
008877-10G
008877-25G
FLUO008877-10GEA
0
EUR
FLUO008877-10G
FLUO008877-25G
3,4-(Methylendioxy)benzylamin
3,4-(Methylendioxy)benzylamin
Formel:
C₈H₉NO₂ Molecular Weight: 151,17 g/mol |
MDL:
MFCD00005840 CAS-Nummer: 2620-50-0 EINECS: 220-056-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...