1-Methyl-4-imidazolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 1-Methyl-1H-imidazole-4-sulphonyl chloride

CFF279.1 CFF279.5 017325-1G 017325-5G
CHMPCFF279.1EA 268 EUR
CHMPCFF279.1 CHMPCFF279.5 FLUO017325-1G FLUO017325-5G
1-Methyl-4-imidazolsulfonylchlorid
1-Methyl-4-imidazolsulfonylchlorid
Formel: C₄H₅ClN₂O₂S
Molecular Weight: 180,61 g/mol
MDL: MFCD00068060
CAS-Nummer: 137049-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR