1-Tritylimidazol-2-carbaldehyd
Lieferant: COMBI-BLOCKS
Synonyme:
1-Trityl-1H-imidazole-2-carbaldehyde
HI-1227-5G
HI-1227-1G
COBBHI-1227-5GEA
569
EUR
COBBHI-1227-5G
COBBHI-1227-1G
1-Tritylimidazol-2-carbaldehyd
1-Tritylimidazol-2-carbaldehyd
Formel:
C₂₃H₁₈N₂O Molecular Weight: 338.400 g/mol |
MDL:
MFCD03453110 CAS-Nummer: 67478-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...