1-Tritylimidazol-4-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-Trityl-1H-imidazole-4-carboxylic acid

HI-1344-1G HI-1344-5G
COBBHI-1344-1GEA 303 EUR
COBBHI-1344-1G COBBHI-1344-5G
1-Tritylimidazol-4-carbonsäure
1-Tritylimidazol-4-carbonsäure
Formel: C₂₃H₁₈N₂O₂
Molecular Weight: 354,4 g/mol
MDL: MFCD06203351
CAS-Nummer: 191103-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR