Dimetridazol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,2-Dimethyl-5-nitroimidazole

L11303.06 L11303.14
L11303.06EA 32.7 EUR
L11303.06 L11303.14
Dimetridazol ≥97%
Dimetridazol

Purity: 97% CAS No.: 551-92-8 Molecular Formula: C5H7N3O2 Formula Weight: 141.13 Melting Point: 136-138° Beilstein Reference: 130665 Merck Reference: 14,3265 MDL No.: MFCD00047046

Formel: C₅H₇N₃O₂
Molecular Weight: 141,13 g/mol
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047046
CAS-Nummer: 551-92-8
EINECS: 209-001-2
Merck Index: 14,3265

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR