Dimetridazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Dimethyl-5-nitroimidazole

M-Clarity™ Qualitätsstufe = MQ200

D4025-100G D4025-1KG
SIALD4025-100GEA 131 EUR
SIALD4025-100G SIALD4025-1KG
Dimetridazol, Sigma-Aldrich®
Dimetridazol
Formel: C₅H₇N₃O₂
Molecular Weight: 141,13 g/mol
MDL: MFCD00047046
CAS-Nummer: 551-92-8
EINECS: 209-001-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR