(1-Methyl-1H-imidazol-2-yl)methanol
Lieferant: BIONET RESEARCH
Synonyme:
1-Methyl-2-imidazolemethanol
12H-013.500
12H-013.1
12H-013.5
BIOT12H-013.500EA
0
EUR
BIOT12H-013.500
BIOT12H-013.1
BIOT12H-013.5
(1-Methyl-1H-imidazol-2-yl)methanol
(1-Methyl-1H-imidazol-2-yl)methanol
Formel:
C₅H₈N₂O Molecular Weight: 112.13 g/mol |
MDL:
MFCD00964673 CAS-Nummer: 17334-08-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...