1,2-Dimethyl-4-imidazolsulfonamid

Lieferant: FLUOROCHEM

CFF317.0.25 CFF317.0.5 CFF317.1 017356-250MG 017356-500MG 017356-1G
CHMPCFF317.0.25EA 427 EUR
CHMPCFF317.0.25 CHMPCFF317.0.5 CHMPCFF317.1 FLUO017356-250MG FLUO017356-500MG FLUO017356-1G
1,2-Dimethyl-4-imidazolsulfonamid
1,2-Dimethyl-4-imidazolsulfonamid
Formel: C₅H₉N₃O₂S
MDL: MFCD02091383
CAS-Nummer: 415913-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR