3-Brom-1-(toluol-4-sulfonyl)indol

Lieferant: COMBI-BLOCKS

IN-8028-5G IN-8028-25G IN-8028-1G
COBBIN-8028-5GEA 181 EUR
COBBIN-8028-5G COBBIN-8028-25G COBBIN-8028-1G
3-Brom-1-(toluol-4-sulfonyl)indol
3-Brom-1-(toluol-4-sulfonyl)indol
Formel: C₁₅H₁₂BrNO₂S
MDL: MFCD09260442
CAS-Nummer: 90481-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR