1-(p-Toluolsulfonyl)indol-3-carbaldehyd
Lieferant: BIONET RESEARCH
Synonyme:
1-Tosyl-1H-indole-3-carbaldehyde
9P-917.500
9P-917.1
9P-917.5
BIOT9P-917.500EA
0
EUR
BIOT9P-917.500
BIOT9P-917.1
BIOT9P-917.5
1-(p-Toluolsulfonyl)indol-3-carbaldehyd
1-(p-Toluolsulfonyl)indol-3-carbaldehyd
Formel:
C₁₅H₁₃NO₂S Molecular Weight: 299,3443 g/mol |
MDL:
MFCD00238944 CAS-Nummer: 50562-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...