1-(p-Toluolsulfonyl)indol 95%
Lieferant: Thermo Scientific
Synonyme:
1-[(4-Methylphenyl)sulphonyl]-1H-indole
B22320.03
B22320.06
B22320.14
B22320.03EA
47.9
EUR
B22320.03
B22320.06
B22320.14
1-(p-Toluolsulfonyl)indol 95%
1-(p-Toluolsulfonyl)indol
Purity: 95%
CAS No.: 31271-90-6
Molecular Formula: C15H13NO2S
Formula Weight: 271.33
Melting Point: 84-86°
MDL No.: MFCD01165032
Formel:
C₁₅H₁₃NO₂S Molecular Weight: 271,33 g/mol Schmelzpunkt: 84…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01165032 CAS-Nummer: 31271-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...