4-(Cyanmethyl)tetrahydropyran

Lieferant: COMBI-BLOCKS

Synonyme: Tetrahydro-2H-pyran-4-ylacetonitrile, 2-(Tetrahydro-2H-pyran-4-yl)acetonitrile

AM-1018-5G AM-1018-1G
COBBAM-1018-5GEA 574 EUR
COBBAM-1018-5G COBBAM-1018-1G
4-(Cyanmethyl)tetrahydropyran
4-(Cyanmethyl)tetrahydropyran
Formel: C₇H₁₁NO
Molecular Weight: 125,1700 g/mol
MDL: MFCD04973733
CAS-Nummer: 850429-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR