1-Boc-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(1-tert-Butoxycarbonyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N-Boc-pyrazole-4-boronic acid pinacol ester, tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(tert-Butoxycarbonyl)pyrazolyl-4-boronic acid Pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-butyl ester, 1-Boc-pyrazolyl-4-boronic acid Pinacol ester, 2-(1-Boc-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-butyl 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-Boc-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylic acid tert-butyl ester, tert-Butyl 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate
M-Clarity™ Qualitätsstufe = MQ100
Formel:
C₁₄H₂₃N₂O₄B Molecular Weight: 294,1544 g/mol |
MDL:
MFCD05663873 CAS-Nummer: 552846-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...