1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole

PN-8782-1G PN-8782-250M
COBBPN-8782-1GEA 510 EUR
COBBPN-8782-1G COBBPN-8782-250M
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester
1-[(2-Trimethylsilylethoxy)methyl]-1H-pyrazol-5-boronsäurepinakolester
Formel: C₁₅H₂₉BN₂O₃Si
Molecular Weight: 324.31 g/mol
MDL: MFCD09037502
CAS-Nummer: 903550-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR