1-(2,2-Diethoxyethyl)-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-6004-1G PN-6004-250M
COBBPN-6004-1GEA 423 EUR
COBBPN-6004-1G COBBPN-6004-250M
1-(2,2-Diethoxyethyl)-1H-pyrazol-4-boronsäurepinakolester
1-(2,2-Diethoxyethyl)-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₅H₂₇BN₂O₄
MDL: MFCD16295107
CAS-Nummer: 1256359-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR