1-Boc-3-methyl-1H-pyrazol-5-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-Boc-3-methyl-1H-pyrazole-5-boronic Acid

BB-6020-5G BB-6020-1G
COBBBB-6020-5GEA 2210 EUR
COBBBB-6020-5G COBBBB-6020-1G
1-Boc-3-methyl-1H-pyrazol-5-boronsäure
1-Boc-3-methyl-1H-pyrazol-5-boronsäure
Formel: C₉H₁₅BN₂O₄
Molecular Weight: 226,04 g/mol
MDL: MFCD15143457
CAS-Nummer: 1217501-27-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR