1-Boc-3,5-dimethyl-1H-pyrazol-4-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(1-Boc-3,5-dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-Butyl ester, 1-Boc-3,5-dimethylpyrazole-4-boronic acid Pinacol ester, tert-Butyl 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3,5-dimethylpyrazole-4-boronic acid Pinacol ester, 2-(1-tert-Butoxycarbonyl-3,5-dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-Butyl 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-Boc-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole

PN-8766-1G PN-8766-250M
COBBPN-8766-1GEA 246 EUR
COBBPN-8766-1G COBBPN-8766-250M
1-Boc-3,5-dimethyl-1H-pyrazol-4-boronsäurepinakolester
1-Boc-3,5-dimethyl-1H-pyrazol-4-boronsäurepinakolester
Formel: C₁₆H₂₇BN₂O₄
Molecular Weight: 322,2076 g/mol
MDL: MFCD09027070
CAS-Nummer: 1073354-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR