1-Boc-3,5-dimethyl-1H-pyrazol-4-boronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(1-Boc-3,5-dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-Butyl ester, 1-Boc-3,5-dimethylpyrazole-4-boronic acid Pinacol ester, tert-Butyl 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3,5-dimethylpyrazole-4-boronic acid Pinacol ester, 2-(1-tert-Butoxycarbonyl-3,5-dimethyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, tert-Butyl 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-Boc-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
M-Clarity™ Qualitätsstufe = MQ100
Formel:
C₁₆H₂₇BN₂O₄ Molecular Weight: 322,2076 g/mol |
MDL:
MFCD09027070 CAS-Nummer: 1073354-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...