(1-(tert-Butoxycarbonyl)-3,5-dimethyl-1H-pyrazol-4-yl)boronic acid

Lieferant: COMBI-BLOCKS

Synonyme: [3,5-Dimethyl-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrazol-4-yl]boronic acid, 1-BOC-3,5-dimethylpyrazole-4-boronic acid

BB-8766-1G BB-8766-250M
COBBBB-8766-1GEA 154 EUR
COBBBB-8766-1G COBBBB-8766-250M
(1-(tert-Butoxycarbonyl)-3,5-dimethyl-1H-pyrazol-4-yl)boronic acid
(1-(tert-Butoxycarbonyl)-3,5-dimethyl-1H-pyrazol-4-yl)boronic acid
Formel: C₁₀H₁₇BN₂O₄
Molecular Weight: 240,06 g/mol
MDL: MFCD12913948
CAS-Nummer: 947533-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR