1-(2,6-Dichlorphenyl)-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

BB-6129-1G BB-6129-250M
COBBBB-6129-1GEA 1630 EUR
COBBBB-6129-1G COBBBB-6129-250M
1-(2,6-Dichlorphenyl)-1H-pyrazol-4-boronsäure
1-(2,6-Dichlorophenyl)-1H-pyrazole-4-boronic acid
Formel: C₉H₇BCl₂N₂O₂
MDL: MFCD15143458
CAS-Nummer: 1217501-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR