1-(2-Chlorphenyl)-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

BB-5120-1G BB-5120-250M
COBBBB-5120-1GEA 1400 EUR
COBBBB-5120-1G COBBBB-5120-250M
1-(2-Chlorphenyl)-1H-pyrazol-4-boronsäure
1-(2-Chlorophenyl)-1H-pyrazole-4-boronic acid
Formel: C₉H₈BClN₂O₂
MDL: MFCD09972101
CAS-Nummer: 1072945-91-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR