1-(4-Fluorphenyl)-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

BB-5119-5G BB-5119-1G
COBBBB-5119-5GEA 1370 EUR
COBBBB-5119-5G COBBBB-5119-1G
1-(4-Fluorphenyl)-1H-pyrazol-4-boronsäure
1-(4-Fluorophenyl)-1H-pyrazole-4-boronic acid
Formel: C₉H₈BFN₂O₂
MDL: MFCD09972100
CAS-Nummer: 1072945-89-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR