2,2'-Bipyridin-3,3'-dicarbonitril

Lieferant: COMBI-BLOCKS

PY-7345-250M PY-7345-100M
COBBPY-7345-250MEA 0 EUR
COBBPY-7345-250M COBBPY-7345-100M
2,2'-Bipyridin-3,3'-dicarbonitril
2,2'-Bipyridin-3,3'-dicarbonitril
Formel: C₁₂H₆N₄
MDL: MFCD09972203
CAS-Nummer: 136869-49-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR