2-(Ethoxycarbonylmethylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-((5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)amino)acetate

PN-5932-1G PN-5932-250M
COBBPN-5932-1GEA 595 EUR
COBBPN-5932-1G COBBPN-5932-250M
2-(Ethoxycarbonylmethylamino)-5-pyrimidinboronsäurepinakolester
2-(Ethoxycarbonylmethylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₄H₂₂BN₃O₄
Molecular Weight: 307,1500 g/mol
MDL: MFCD12546563
CAS-Nummer: 1202805-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR