2-(Isopropylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: N-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

PN-5923-1G PN-5923-250M
COBBPN-5923-1GEA 410 EUR
COBBPN-5923-1G COBBPN-5923-250M
2-(Isopropylamino)-5-pyrimidinboronsäurepinakolester
2-(Isopropylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₃H₂₂BN₃O₂
Molecular Weight: 263,1400 g/mol
MDL: MFCD12546556
CAS-Nummer: 1218791-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR