2-(Ethylamino)-5-pyrimidinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
N-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
PN-5921-1G
PN-5921-250M
COBBPN-5921-1GEA
225
EUR
COBBPN-5921-1G
COBBPN-5921-250M
2-(Ethylamino)-5-pyrimidinboronsäurepinakolester
2-(Ethylamino)-5-pyrimidinboronsäurepinakolester
Formel:
C₁₂H₂₀BN₃O₂ Molecular Weight: 249,1200 g/mol |
MDL:
MFCD12546554 CAS-Nummer: 1218791-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...