2-(Methylsulfonylamino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5936-1G PN-5936-250M
COBBPN-5936-1GEA 1490 EUR
COBBPN-5936-1G COBBPN-5936-250M
2-(Methylsulfonylamino)-5-pyrimidinboronsäurepinakolester
2-(Methylsulfonylamino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₁H₁₈BN₃O₄S
MDL: MFCD12546566
CAS-Nummer: 1218789-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR