2-(Methylthio)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Methylthio)pyrimidine-5-boronic acid pinacol ester, 2-(Methylthio)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

PN-8720-5G PN-8720-1G
COBBPN-8720-5GEA 497 EUR
COBBPN-8720-5G COBBPN-8720-1G
2-(Methylthio)-5-pyrimidinboronsäurepinakolester
2-(Methylthio)-5-pyrimidinboronsäurepinakolester
Formel: C₁₁H₁₇BN₂O₂S
Molecular Weight: 252.140 g/mol
MDL: MFCD07781247
CAS-Nummer: 940284-18-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR