2-(Boc-amino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Aminopyrimidine-5-boronic acid, pinacol ester, 2-BOC protected

PN-8765-1G PN-8765-250M
COBBPN-8765-1GEA 324 EUR
COBBPN-8765-1G COBBPN-8765-250M
2-(Boc-amino)-5-pyrimidinboronsäurepinakolester
2-(Boc-amino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₅H₂₄BN₃O₄
MDL: MFCD09027069
CAS-Nummer: 1032758-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR