2-(Methylthio)-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Methylthio)pyrimidin-5-yl)boronic acid, 2-(Methylthio)pyrimidine-5-boronic acid

BB-9028-5G BB-9028-1G
COBBBB-9028-5GEA 761 EUR
COBBBB-9028-5G COBBBB-9028-1G
2-(Methylthio)-5-pyrimidinboronsäure
2-(Methylthio)-5-pyrimidineboronic acid
Formel: C₅H₇BN₂O₂S
Molecular Weight: 170,0000 g/mol
MDL: MFCD07375142
CAS-Nummer: 348098-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR