4,6-Dichlor-2-(methylthio)pyrimidin

Lieferant: CHEMPUR

SPB07199.250 SPB07199.1
CHMPSPB07199.250EA 0 EUR
CHMPSPB07199.250 MAYBSPB07199.1
4,6-Dichlor-2-(methylthio)pyrimidin
4,6-Dichloro-2-(methylthio)pyrimidine
Formel: C₅H₄Cl₂N₂S
Molecular Weight: 195.07 g/mol
MDL: MFCD00006086
CAS-Nummer: 6299-25-8
EINECS: 228-577-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR