4-Methyl-2-(methylthio)pyrimidin

Lieferant: BIONET RESEARCH

Synonyme: 4-Methyl-2(methylsulphanyl)pyrimidine

6Y-0605.1 6Y-0605.10 6Y-0605.5 6Y-0605.500
BIOT6Y-0605.1EA 0 EUR
BIOT6Y-0605.1 BIOT6Y-0605.10 BIOT6Y-0605.5 BIOT6Y-0605.500
4-Methyl-2-(methylthio)pyrimidin
4-Methyl-2-(methylthio)pyrimidine
Formel: C₆H₈N₂S
Molecular Weight: 140.21 g/mol
MDL: MFCD00023242
CAS-Nummer: 14001-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR