2-(Cbz-amino)-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-6296-1G PN-6296-250M
COBBPN-6296-1GEA 618 EUR
COBBPN-6296-1G COBBPN-6296-250M
2-(Cbz-amino)-5-pyrimidinboronsäurepinakolester
2-(Cbz-amino)-5-pyrimidinboronsäurepinakolester
Formel: C₁₈H₂₂BN₃O₄
MDL: MFCD16295114
CAS-Nummer: 1256359-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR