2-Benzyloxy-5-pyrimidinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

PN-5859-1G PN-5859-250M
COBBPN-5859-1GEA 669 EUR
COBBPN-5859-1G COBBPN-5859-250M
2-Benzyloxy-5-pyrimidinboronsäurepinakolester
2-Benzyloxy-5-pyrimidinboronsäurepinakolester
Formel: C₁₇H₂₁BN₂O₃
Molecular Weight: 312.1700 g/mol
MDL: MFCD11878343
CAS-Nummer: 1218791-34-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR