2,4-Dibenzyloxy-5-pyrimidinboronsäureneopentylglycolester
Lieferant: COMBI-BLOCKS
NE-2243-250M
NE-2243-100M
COBBNE-2243-250MEA
0
EUR
COBBNE-2243-250M
COBBNE-2243-100M
2,4-Dibenzyloxy-5-pyrimidinboronsäureneopentylglycolester
2,4-Dibenzyloxy-5-pyrimidinboronsäureneopentylglycolester
Formel:
C₂₃H₂₅BN₂O₄ |
MDL:
MFCD09037496 CAS-Nummer: 1072944-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...