2-(4'-Methoxybenzyloxy)-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS

BB-5813-1G BB-5813-250M
COBBBB-5813-1GEA 1630 EUR
COBBBB-5813-1G COBBBB-5813-250M
2-(4'-Methoxybenzyloxy)-5-pyrimidinboronsäure
2-(4'-Methoxybenzyloxy)-5-pyrimidinboronsäure
Formel: C₁₂H₁₃BN₂O₄
MDL: MFCD12546478
CAS-Nummer: 1217500-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR