Ethylpyrrol-2-carboxylat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Ethyl 1H-pyrrole-2-carboxylate

L16382.06 L16382.14
L16382.06EA 85.4 EUR
L16382.06 L16382.14
Ethylpyrrol-2-carboxylat ≥98%
Ethylpyrrol-2-carboxylat

Purity: 98+% CAS No.: 2199-43-1 Molecular Formula: C7H9NO2 Formula Weight: 139.16 Melting Point: 37-39° Boiling Point: 70°/0.8mm MDL No.: MFCD00817049

Formel: C₇H₉NO₂
Molecular Weight: 139.16 g/mol
Siedepunkt: 70 °C (0.8 mmHg)
Schmelzpunkt: 37…39 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00817049
CAS-Nummer: 2199-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR