Ethylpyrrol-2-carboxylat

Lieferant: BIONET RESEARCH

Synonyme: Ethyl 1H-pyrrole-2-carboxylate

3X-0740.10 3X-0740.500 3X-0740.1 3X-0740.5
BIOT3X-0740.10EA 0 EUR
BIOT3X-0740.10 BIOT3X-0740.500 BIOT3X-0740.1 BIOT3X-0740.5
Ethylpyrrol-2-carboxylat
Ethylpyrrol-2-carboxylat
Formel: C₇H₉NO₂
Molecular Weight: 139.16 g/mol
MDL: MFCD00817049
CAS-Nummer: 2199-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR