Ethylpyrrol-2-carboxylat

Lieferant: CHEMPUR

Synonyme: Ethyl 1H-pyrrole-2-carboxylate

SEW05579.250 SEW05579.1
CHMPSEW05579.250EA 0 EUR
CHMPSEW05579.250 MAYBSEW05579.1
Ethylpyrrol-2-carboxylat
Ethylpyrrol-2-carboxylat
Formel: C₇H₉NO₂
Molecular Weight: 139,16 g/mol
MDL: MFCD00817049
CAS-Nummer: 2199-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR