4-Brom-2-(trichloracetyl)pyrrol

Lieferant: COMBI-BLOCKS

Synonyme: 1-(4-Bromo-1H-pyrrol-2-yl)-2,2,2-trichloroethanone

HC-2131-5G HC-2131-1G
COBBHC-2131-5GEA 353 EUR
COBBHC-2131-5G COBBHC-2131-1G
4-Brom-2-(trichloracetyl)pyrrol
4-Brom-2-(trichloracetyl)pyrrol
Formel: C₆H₃BrCl₃NO
Molecular Weight: 291,3571 g/mol
MDL: MFCD00832836
CAS-Nummer: 72652-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR