1-Methylpyrrol-2-carbonsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Methyl-1H-pyrrole-2-carboxylic acid, 1-Methyl-2-pyrrolecarboxylic acid

L05906.03 L05906.06
L05906.03EA 26.7 EUR
L05906.03 L05906.06
1-Methylpyrrol-2-carbonsäure ≥98%
1-Methylpyrrole-2-carboxylic acid

Purity: 98% CAS No.: 6973-60-0 Molecular Formula: C6H7NO2 Formula Weight: 125.13 Melting Point: 135-138° Beilstein Reference: 115703

Formel: C₆H₇NO₂
Molecular Weight: 125,13 g/mol
Schmelzpunkt: 135…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003088
CAS-Nummer: 6973-60-0
EINECS: 230-217-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR